Anti-IL17A Rat Monoclonal Antibody (Unconjugated) [clone: 20B4.G10.F5]

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

210-501-B32
ROCK210-501-B32EA 841 EUR
ROCK210-501-B32
Anti-IL17A Rat Monoclonal Antibody (Unconjugated) [clone: 20B4.G10.F5]
Antilichamen
IL-17A monoclonal antibody has been tested for use in western blotting, flow cytometry and ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 23 kDa in size.

Recommended Dilutions: ELISA: 1:10000; Flow Cytometry: 1:500; Western Blot: 1:1000;

Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: 20B4.G10.F5
Conjugation: Unconjugated
Epitope: Full length
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR