Anti-TUBB3 Mouse Polyclonal Antibody (Unconjugated) [clone: AA10]

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

212-301-E55
ROCK212-301-E55EA 976 EUR
ROCK212-301-E55
Anti-TUBB3 Mouse Polyclonal Antibody (Unconjugated) [clone: AA10]
Antilichamen
Type: Primary
Antigen: TUBB3
Clonality: Polyclonal
Clone: AA10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR