Anti-IgG F(ab')2 Rabbit Polyclonal Antibody

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

213-4104
ROCK213-4104EA 971 EUR
ROCK213-4104
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
Antilichamen
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR