Anti-H5N1 Mouse Monoclonal Antibody (Unconjugated) [clone: 8A3]

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

500-301-978
ROCK500-301-978EA 841 EUR
ROCK500-301-978
Anti-H5N1 Mouse Monoclonal Antibody (Unconjugated) [clone: 8A3]
Antilichamen
This monoclonal antibody can be used for hemagglutination inhibition (HI) assays to provide antigenic characterization of the influenza A viruses of the H5 HA subtype.

Recommended Dilutions: ELISA: 1:5000;

Type: Primary
Antigen: H5N1
Clonality: Monoclonal
Clone: 8A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Virus, H5N1 Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR