Anti-Biotin Rabbit Polyclonal Antibody

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

600-401-098
ROCK600-401-098EA 566 EUR
ROCK600-401-098
Anti-Biotin Rabbit Polyclonal Antibody
Antilichamen
Suitable for immunoprecipitation, immunodiffusion, conjugation and most immunological methods requiring lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:200,000; Western Blot: 1:2000 - 1:10000;

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Specificaties

Environmentally Preferable
Type antilichaam Label,Biotin
Type antilichaam Label,Biotin
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemie Yes
ImmunoNeerslag Yes
Immunogeen Biotin conjugated to Keyhole Limpet Hemocyanin (KLH)
Formaat Antibody fraction
Formulier Liquid (sterile filtered)
Purification purified by immunoaffinity chromatography
Kruisadsorptie No
Opslagbuffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
Opslagtemperatuur –20 °C
Shipping temperature Wet Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR