Anti-FANCC Rabbit Polyclonal Antibody

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

600-401-672
ROCK600-401-672EA 821 EUR
ROCK600-401-672
Anti-FANCC Rabbit Polyclonal Antibody
Antilichamen
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 63 kDa in size corresponding to FANCC by western blotting.

Recommended Dilutions: ELISA: 1:20000 - 1:80,000; Western Blot: 1:1000 - 1:3000;

Type: Primary
Antigen: FANCC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR