Anti-RAF1 Rabbit Polyclonal Antibody

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

600-401-E23
ROCK600-401-E23EA 976 EUR
ROCK600-401-E23
Anti-RAF1 Rabbit Polyclonal Antibody
Antilichamen
Anti-Raf-1 pS301antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 74 kDa in size corresponding to the Raf-1 Protein phosphorylated at Ser301.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR