Anti-SMAD3 Rabbit Polyclonal Antibody

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

600-401-E70S
ROCK600-401-E70SEA 226 EUR
ROCK600-401-E70S
Anti-SMAD3 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: ELISA: 1:15000-1:75000; Western Blot: 1:1000; Other Dilution: User Optimized

Type: Primary
Antigen: SMAD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR