Anti-ACK Rabbit Polyclonal Antibody (Biotin)

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

600-406-939
ROCK600-406-939EA 861 EUR
ROCK600-406-939
Anti-ACK Rabbit Polyclonal Antibody (Biotin)
Antilichamen
This affinity-purified antibody is suitable for use in ELISA and western blotting assays.  The antibody reacts specifically with acetylated lysine residues.

Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:1000-1:5000;

Type: Primary
Antigen: ACK
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mammalian, Avian,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR