Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Texas Red®)

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

603-4904
ROCK603-4904EA 384 EUR
ROCK603-4904
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Texas Red®)
Antilichamen
Secondary Rabbit Anti-IgY F(ab')2 Reacts with Chicken

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR