Anti-IgG (H+L) Goat Polyclonal Antibody [clone: 10H11.E12] (DyLight® 488)

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

610-141-121
ROCK610-141-121EA 216 EUR
ROCK610-141-121
Anti-IgG (H+L) Goat Polyclonal Antibody [clone: 10H11.E12] (DyLight® 488)
Antilichamen
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone: 9F9.F9
Conjugation: DyLight® 488
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Specificaties

antigeen symbool IgG (H+L)
Environmentally Preferable
Type antilichaam Ondergeschikt
Clonality Polyklonaal
Conjugatie DyLight® 488
Clone 10H11.E12
Reactivity Mouse
Host Goat
Isotype IgG
Western blot Yes
ImmunoFluorescentie Yes
Immunogeen Mouse IgG whole molecule
Formaat Antibody fraction
Formulier Lyophilized
Purification purified by immunoaffinity chromatography
Kruisadsorptie No
Opslagbuffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Opslagtemperatuur –20 °C
Shipping temperature Ambient
Tested applications Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR