Anti-IgG F(ab')2 Goat Polyclonal Antibody

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

611-1104
ROCK611-1104EA 906 EUR
ROCK611-1104
Anti-IgG F(ab')2 Goat Polyclonal Antibody
Antilichamen
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Order Now

Specificaties

antigeen symbool IgG F(ab')2
Environmentally Preferable
Type antilichaam Ondergeschikt
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Rabbit
Host Goat
Isotype
Western blot Yes
ELISA Yes
ImmunoChemie Yes
ImmunoNeerslag Yes
ImmunoFluorescentie Yes
Gelverschuiving Yes
Flowcytometrie Yes
ChIP Yes
Neutralisation Yes
Kruisadsorptie none
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR