Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

620-105-440
ROCK620-105-440EA 553 EUR
ROCK620-105-440
Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antilichamen
Recommended Dilutions: ELISA: 1:2000-1:10000; Western Blot: 1:500-1:3000; Immunochemistry: 1:200-1:1000
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Armenian, Golden Syrian Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR