Anti-IgG (H+L) Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

704-405-002
ROCK704-405-002EA 421 EUR
ROCK704-405-002
Anti-IgG (H+L) Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antilichamen
This product has been assayed against 1.0 µg of Dog IgG in a standard capture ELISA using pNPP p-nitrophenyl phosphate code # NPP-10 as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Dog
Order Now

Specificaties

antigeen symbool IgG (H+L)
Environmentally Preferable
Type antilichaam Ondergeschikt
Clonality Polyklonaal
Conjugatie AP (Alkaline Phosphatase)
Reactivity Dog
Host Rabbit
Isotype
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Antilichaamfragment F(ab')2
Immunogeen Dog IgG whole molecule
Formaat Antibody fraction
Formulier Liquid (sterile filtered)
Purification purified by immunoaffinity chromatography
Kruisadsorptie No
Opslagbuffer 0,05 M Tris Chloride, 0,15M Sodium Chloride, 0,001M Magnesium Chloride, 0,0001M Zinc Chloride, 50% (v/v) Glycerol
Opslagtemperatuur +4 °C
Shipping temperature Wet Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR