Anti-IgG (H+L) Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

705-703-125
ROCK705-703-125EA 436 EUR
ROCK705-703-125
Anti-IgG (H+L) Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:500 - 1:5000;
Restore with deionized water (or equivalent); 500 µl
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR