Anti-IgG (H+L) Goat Polyclonal Antibody

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

710-1132
ROCK710-1132EA 366 EUR
ROCK710-1132
Anti-IgG (H+L) Goat Polyclonal Antibody
Antilichamen
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR