Anti-IgG (H+L) Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

711-1222
ROCK711-1222EA 341 EUR
ROCK711-1222
Anti-IgG (H+L) Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antilichamen
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Specificaties

antigeen symbool IgG (H+L)
Environmentally Preferable
Type antilichaam Ondergeschikt
Clonality Polyklonaal
Conjugatie FITC (Fluoresceïne-isothiocyanaat)
Reactivity Rabbit
Host Goat
Isotype
Western blot Yes
ELISA Yes
ImmunoChemie Yes
ImmunoNeerslag Yes
ImmunoFluorescentie Yes
Gelverschuiving Yes
Flowcytometrie Yes
ChIP Yes
Neutralisation Yes
Antilichaamfragment F(ab')2
Immunogeen Rabbit IgG whole molecule
Formaat Antibody fraction
Formulier Lyophilized
Purification purified by immunoaffinity chromatography
Concentratie 1,0 mg/ml by UV absorbance at 280 nm
Opslagbuffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR