Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

807-1302
ROCK807-1302EA 436 EUR
ROCK807-1302
Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)
Antilichamen
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Golden Syrian Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR