3,3'-Diaminobenzidine membrane peroxidase substrate (50X)

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

DAB-10
ROCKDAB-10EA 290 EUR
ROCKDAB-10
3,3'-Diaminobenzidine membrane peroxidase substrate (50X)
3,3'-Diaminobenzidine

DAB membrane peroxidase substrate (50X) is a benzidine derivative oxidized by hydrogen peroxide in the presence of hemoglobin yielding a dark-brown colour of solution. This oxidation can be utilized for tissue samples by first preparing them with hydrogen peroxide.


Suitable for use in Western blotting, immunohistochemistry or other immunology, and microbiology research.


Bestelinformatie: Diluent buffer is not included and has to be ordered separately.


Opgelet: Store at 4 °C prior to opening and protect from moisture and light.

Formule: C₁₂H₁₄N₄
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007725
CAS nummer: 91-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR