Lysates, human
Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
W09-000-365 W09-000-361 W09-000-362 W09-000-364 W09-000-363 W09-001-370 W09-000-366 W09-000-360 W09-001-GY4
ROCKW09-000-365EA 165 EUR
ROCKW09-000-365 ROCKW09-000-361 ROCKW09-000-362 ROCKW09-000-364 ROCKW09-000-363 ROCKW09-001-370 ROCKW09-000-366 ROCKW09-000-360 ROCKW09-001-GY4
Lysates, human
Cellen
Whole cell lysates are derived from cell lines tissues using highly refined extraction protocols to ensure quality, protein integrity and high reproducibility. Lysates are prepared in denaturing buffer without dissociating agents.
  • Liquid
Whole cell lysates are ready to use for SDS-PAGE and Western blotting purposes. All extracts are tested by SDS-PAGE using 4 to 20% gradient gels and immunoblot analysis using antibodies to key cell signaling components to confirm the presence of both high molecular weight and low molecular weight proteins.
Each vial contains 1mg/ml of protein in SDS-PAGE sample buffer. The sample buffer consists of 0,05 mM Tris Hcl, 2% SDS, 10% glycerol and 0,1% bromophenol blue at pH 6,8.