N,N-Diethyl-1,1,1-trimethylsilylamine

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Trimethylsilyldiethylamine, (Diethylamino)trimethylsilane, N,N-Diethylaminotrimethylsilane, N,N-Diethyltrimethylsilylamine, N-(Trimethylsilyl)diethylamine, TMSDEA

0013.1
ROTH0013.1EA 165 EUR
ROTH0013.1
N,N-Diethyl-1,1,1-trimethylsilylamine
N,N-Diethyl-1,1,1-trimethylsilylamine
Formule: C₇H₁₉NSi
MW: 145.32 g/mol
Kookpunt: 126…127 °C (1013 hPa)
Smeltpunt: –10 °C
Dichtheid: 0.7074 g/cm³ (20 °C)
Vlampunt: 15
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009040
CAS nummer: 996-50-9
EINECS: 213-637-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR