Nystatin dihydrate, 4.400 U/mg

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0241.1 0241.2
ROTH0241.1EA 0 EUR
ROTH0241.1 ROTH0241.2
Nystatin dihydrate, 4.400 U/mg
Nystatin dihydrate
Formule: C₄₇H₇₅NO₁₇·2H₂O
MDL Nummer: MFCD00036240
CAS nummer: 1400-61-9
EINECS: 215-749-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR