Magnesiumacetaat tetrahydraat

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate

0275.1 0275.2 0275.3
ROTH0275.1EA 0 EUR
ROTH0275.1 ROTH0275.2 ROTH0275.3
Magnesiumacetaat tetrahydraat
Magnesium acetate tetrahydrate
Formule: (H₃CCOO)₂Mg·4H₂O
MW: 214.46 g/mol
Smeltpunt: 80
Dichtheid: 1.454 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149214
CAS nummer: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR