Auramine O

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

0319.1 0319.2
ROTH0319.1EA 0 EUR
ROTH0319.1 ROTH0319.2
Auramine O
Auramine O
Formule: C₁₇H₂₂ClN₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012484
CAS nummer: 2465-27-2
EINECS: 219-567-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR