2-Butoxyethanol

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Ethylene glycol butyl ether, Butyl cellosolve, Ethylene glycol monobutyl ether

0341.1 0341.2
ROTH0341.1EA 0 EUR
ROTH0341.1 ROTH0341.2
2-Butoxyethanol
2-Butoxyethanol
Formule: CH₃(CH₂)₃OCH₂CH₂OH
MW: 118.18 g/mol
Kookpunt: 171
Smeltpunt: –70 °C
Dichtheid: 0.902 g/cm³ (20 °C)
Vlampunt: 50
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002884
CAS nummer: 111-76-2
EINECS: 203-905-0
UN: 2810
ADR: 6.1 III
Merck Index: 13,01558
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5a9d2c527d0c167d2248d1936e2d1bd4aeab7a&vid=75260b2608ddc5eb29642178ab193fbcaf54198d&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR