Alizarin red S (sodium salt)

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

0348.1 0348.2
ROTH0348.1EA 0 EUR
ROTH0348.1 ROTH0348.2
Alizarin red S (sodium salt)
Alizarin red S (sodium salt)
Formule: C₁₄H₇NaO₇S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013049
CAS nummer: 130-22-3
EINECS: 204-981-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR