Acetonitril

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

2259.1
ROTH2259.1EA 0 EUR
ROTH2259.1
Acetonitril
Acetonitril
Formule: H₃CCN
MW: 41.05 g/mol
Kookpunt: 81.6
Smeltpunt: –45.7 °C
Dichtheid: 0.782 g/cm³ (20 °C)
Vlampunt: 2 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001878
CAS nummer: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
Bereik: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR