IPTG (isopropyl-β-D-thiogalactopyranoside)

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

2316.3 2316.1 2316.2 2316.4 2316.5
ROTH2316.3EA 0 EUR
ROTH2316.3 ROTH2316.1 ROTH2316.2 ROTH2316.4 ROTH2316.5
IPTG (isopropyl-β-D-thiogalactopyranoside)
IPTG (isopropyl-β-D-thiogalactopyranoside)
Formule: C₉H₁₈O₅S
Kookpunt: 402 °C (1013 hPa)
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00063273
CAS nummer: 367-93-1
EINECS: 206-703-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR