Adenosine

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

2322.3
ROTH2322.3EA 0 EUR
ROTH2322.3
Adenosine
Adenosine
Formule: C₁₀H₁₃N₅O₄
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005752
CAS nummer: 58-61-7
EINECS: 200-389-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR