N,N,N',N'-Tetramethylethyleendiamine

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

2367.1 2367.2 2367.3
ROTH2367.1EA 0 EUR
ROTH2367.1 ROTH2367.2 ROTH2367.3
N,N,N',N'-Tetramethylethyleendiamine
N,N,N',N'-Tetramethylethyleendiamine
Formule: C₆H₁₆N₂
Kookpunt: 120 °C (1013 hPa)
Smeltpunt: –64 °C
Dichtheid: 0,7765 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008335
CAS nummer: 110-18-9
EINECS: 203-744-6
UN: 2372
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6838f1f5b7d0f69dcc81a8ed247f6c178dad231c&vid=ea361367e9944e4101e38622fdd18b07756eb676&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR