Lanthanum(III) oxide

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

3045.1
ROTH3045.1EA 0 EUR
ROTH3045.1
Lanthanum(III) oxide
Lanthanum(III) oxide
Formule: La₂O₃
MW: 325.81 g/mol
Kookpunt: 4200
Smeltpunt: 2315
Dichtheid: 6.51 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011071
CAS nummer: 1312-81-8
EINECS: 215-200-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR