Azulene

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

3159.1
ROTH3159.1EA 0 EUR
ROTH3159.1
Azulene
Azulene
Formule: C₁₀H₈
Kookpunt: 242 °C (1013 hPa)
Smeltpunt: 96…97 °C
MDL Nummer: MFCD00003810
CAS nummer: 275-51-4
EINECS: 205-993-6
UN: 3077
ADR: 9,III
Merck Index: 13,00928

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR