α-Linolenic acid ethyl ester

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

3193.1
ROTH3193.1EA 0 EUR
ROTH3193.1
α-Linolenic acid ethyl ester
α-Linolenic acid ethyl ester
Formule: C₂₀H₃₄O₂
Kookpunt: 218 °C (1013 hPa)
Dichtheid: 0,8919 g/cm³ (20 °C)
MDL Nummer: MFCD07371452
CAS nummer: 1191-41-9
EINECS: 214-734-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR