Vitamin K1 (phytomenadione, phylloquinone)

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

3804.1 3804.2
ROTH3804.1EA 0 EUR
ROTH3804.1 ROTH3804.2
Vitamin K1 (phytomenadione, phylloquinone)
Vitamin K1 (phytomenadione, phylloquinone)
Formule: C₃₁H₄₆O₂
MDL Nummer: MFCD00214063
CAS nummer: 84-80-0
EINECS: 201-564-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR