Citric acid monohydrate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

3958.1 3958.2 3958.3 3958.4 3958.5
ROTH3958.1EA 0 EUR
ROTH3958.1 ROTH3958.2 ROTH3958.3 ROTH3958.4 ROTH3958.5
Citric acid monohydrate
Citric acid monohydrate
Formule: C₆H₈O₇.H₂O
MW: 210.14 g/mol
Smeltpunt: 135…152 °C
Dichtheid: 1.552 g/cm³ (20 °C)
Vlampunt: 173.9 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149972
CAS nummer: 5949-29-1
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR