Lithium hydroxide monohydrate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

3997.2 3997.1
ROTH3997.2EA 0 EUR
ROTH3997.2 ROTH3997.1
Lithium hydroxide monohydrate
Lithium hydroxide monohydrate
Formule: LiOH·1H₂O
Kookpunt: 920 °C (1013 hPa)
Smeltpunt: 462 °C
Dichtheid: 1,51 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149772
CAS nummer: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: 8,II
Merck Index: 13,05556

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR