Mitomycin C

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Mitomycin, Ametycin

4150.1
ROTH4150.1EA 0 EUR
ROTH4150.1
Mitomycin C
Mitomycin C
Formule: C₁₅H₁₈N₄O₅
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00078109
CAS nummer: 50-07-7
EINECS: 200-008-6
UN: 2811
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR