Nitrilotriacetic acid

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: NTA, 2,2',2''-Nitrilotriacetic acid

4384.1
ROTH4384.1EA 0 EUR
ROTH4384.1
Nitrilotriacetic acid
Nitrilotriacetic acid
Formule: C₆H₉NO₆
MW: 191.14 g/mol
Kookpunt: 498
Smeltpunt: 242
Dichtheid: 1.67 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004287
CAS nummer: 139-13-9
EINECS: 205-355-7
UN: 2923
ADR: 8 II
Merck Index: 12,06675

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR