L(+)-Rhamnose monohydrate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal hydrate, 6-Deoxy-L(+)-mannose monohydrate

4655.1 4655.2
ROTH4655.1EA 0 EUR
ROTH4655.1 ROTH4655.2
L(+)-Rhamnose monohydrate
L(+)-Rhamnose monohydrate
Formule: C₆H₁₂O₅·1H₂O
MW: 182.17 g/mol
Smeltpunt: 85…95 °C
Dichtheid: 1.47 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149363
CAS nummer: 10030-85-0
EINECS: 222-793-4
Merck Index: 12,08338

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR