Tributyrin (glycerol tributyrate)

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Tributyrin, Glyceryl tributyrate, Glycerol tributyrate, Propane-1,2,3-triyl tributyrate

4813.1
ROTH4813.1EA 0 EUR
ROTH4813.1
Tributyrin (glycerol tributyrate)
Tributyrin (glycerol tributyrate)
Formule: C₁₅H₂₆O₆
MW: 302.37 g/mol
Kookpunt: 305
Smeltpunt: –70 °C
Dichtheid: 1.035 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009392
CAS nummer: 60-01-5
EINECS: 200-451-5
Merck Index: 12,09750

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR