Ammonium hydrogen sulphate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

5485.1 5485.3
ROTH5485.1EA 0 EUR
ROTH5485.1 ROTH5485.3
Ammonium hydrogen sulphate
Ammonium hydrogen sulphate
Formule: H₅NO₄S
Kookpunt: 350 °C (1013 hPa)
Dichtheid: 1,787 g/cm³ (20 °C)
MDL Nummer: MFCD00051073
CAS nummer: 7803-63-6
EINECS: 232-265-5
UN: 2506
ADR: 8,II
Merck Index: 12,00526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR