Charcoal activated

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Carbon activated, Activated carbon, Activated charcoal

5966.1 5966.2 5966.4
ROTH5966.1EA 0 EUR
ROTH5966.1 ROTH5966.2 ROTH5966.4
Charcoal activated
Charcoal activated
Formule: C
Smeltpunt: 3550 °C
Dichtheid: 2 g/cm³ (20 °C)
MDL Nummer: MFCD00133992
CAS nummer: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
Bereik: 01-2119488894-16
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR