Toluene-α-sulphonyl fluoride

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

6367.1 6367.2 6367.3 6367.4
ROTH6367.1EA 0 EUR
ROTH6367.1 ROTH6367.2 ROTH6367.3 ROTH6367.4
Toluene-α-sulphonyl fluoride
Toluene-α-sulphonyl fluoride
Formule: C₇H₇FO₂S
Kookpunt: 80…85 °C (0,3 mmHg)
Smeltpunt: 91…92 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007424
CAS nummer: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR