Potassium iodide

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

6750.1 6750.2
ROTH6750.1EA 0 EUR
ROTH6750.1 ROTH6750.2
Potassium iodide
Potassium iodide
Formule: Kl
Kookpunt: 1330 °C (1013 hPa)
Smeltpunt: 686 °C
Dichtheid: 3,13 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011405
CAS nummer: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR