di-Potassium hydrogen phosphate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

6875.1 6875.2
ROTH6875.1EA 0 EUR
ROTH6875.1 ROTH6875.2
di-Potassium hydrogen phosphate
di-Potassium hydrogen phosphate
Formule: K₂HPO₄
Smeltpunt: 340 °C
Dichtheid: 2,3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011383
CAS nummer: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR