Dimethylsulfoxide

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

7029.1 7029.2 7029.3
ROTH7029.1EA 69.9 EUR
ROTH7029.1 ROTH7029.2 ROTH7029.3
Dimethylsulfoxide
Dimethylsulfoxide
Formule: (CH₃)₂SO
MW: 78.14 g/mol
Kookpunt: 189
Smeltpunt: 18.5
Dichtheid: 1.101 g/cm³ (20 °C)
Vlampunt: 87
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002089
CAS nummer: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR