DIPSO

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

7151.1
ROTH7151.1EA 0 EUR
ROTH7151.1
DIPSO
DIPSO
Formule: C₇H₁₇NO₆S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00038353
CAS nummer: 68399-80-4
EINECS: 269-992-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR