p-Anisaldehyde

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

7201.1
ROTH7201.1EA 0 EUR
ROTH7201.1
p-Anisaldehyde
p-Anisaldehyde
Formule: C₈H₈O₂
Kookpunt: 140 °C (1013 hPa)
Smeltpunt: 0…2 °C
Dichtheid: 1,12 g/cm³ (25 °C)
Vlampunt: 110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003385
CAS nummer: 123-11-5
EINECS: 204-602-6
Merck Index: 13,00666

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR