Caprinaldehyde

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

7547.1
ROTH7547.1EA 0 EUR
ROTH7547.1
Caprinaldehyde
Caprinaldehyde
Formule: C₁₀H₂₀O
Kookpunt: 207 °C (1013 hPa)
Smeltpunt: 7 °C
Dichtheid: 0,83 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007031
CAS nummer: 112-31-2
EINECS: 203-957-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR