Ureum

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

7638.1 7638.2 7638.5
ROTH7638.1EA 0 EUR
ROTH7638.1 ROTH7638.2 ROTH7638.5
Ureum
Ureum
Formule: NH₂CONH₂
Kookpunt: 197 °C (1013 hPa)
Dichtheid: 1,34 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008022
CAS nummer: 57-13-6
EINECS: 200-315-5
Bereik: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR